Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC030499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287206.1 | NP_001274135.1 | 5, 6 | 561 | ||
GenBank mRNA | AK138440.1 | - | 5, 6 | 553 | ||
BC037104.1 | - | 5, 6 | 499 | |||
BC048862.1 | - | 511 | ||||
BC059191.1 | - | 5, 6 | 510 |