Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085500.2 | NP_001078969.2 | 3 | 662 | ||
GenBank mRNA | BC056217.1 | - | 3 | 657 |