Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50177
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
230
6921326
FQ211494.1
-
230
FQ211970.1
-
226
FQ212357.1
-
227
FQ212579.1
-
226
FQ212640.1
-
226
FQ212691.1
-
226
FQ212789.1
-
230
FQ212869.1
-
230
FQ213028.1
-
226
FQ213029.1
-
224
FQ213091.1
-
226
FQ213115.1
-
232
FQ213200.1
-
229
FQ213216.1
-
226
FQ213310.1
-
119
FQ213366.1
-
225
FQ213496.1
-
229
FQ213520.1
-
230
FQ213526.1
-
226
FQ213581.1
-
226
FQ213598.1
-
226
FQ213637.1
-
226
FQ213703.1
-
226
FQ213760.1
-
226
FQ213762.1
-
231
FQ213913.1
-
226
FQ213941.1
-
230
FQ213981.1
-
227
FQ213985.1
-
120
FQ213999.1
-
225
FQ214059.1
-
230
FQ214181.1
-
230
FQ214262.1
-
230
FQ214491.1
-
226
FQ214500.1
-
226
FQ214519.1
-
226
FQ214525.1
-
226
FQ214540.1
-
230
FQ215159.1
-
230
FQ215317.1
-
226
FQ215600.1
-
230
FQ216397.1
-
226
FQ216471.1
-
227
FQ218091.1
-
226
FQ220250.1
-
226
FQ220325.1
-
226
FQ220698.1
-
230
FQ220779.1
-
226
FQ222184.1
-
229
FQ223357.1
-
230
FQ225272.1
-
226
FQ226994.1
-
230
FQ229228.1
-
226
FQ229580.1
-
230
FQ229619.1
-
226
FQ229905.1
-
225
FQ231022.1
-
230
FQ231702.1
-
230
FQ233337.1
-
229
FQ234377.1
-
230
FQ235266.1
-
227
M17069.1
-
228
M19312.1
-
209
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007589, BY095488, BC100301, L31642, AK013068, and others.
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline