Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080934.1 | NP_001074403.1 | 4 | 2126 | ||
GenBank mRNA | AK037416.1 | - | 3 | 2238 | ||
AK079757.1 | - | 2 | 1171 | |||
BC052932.1 | - | 3 | 1159 | |||
BC053982.1 | - | 3 | 1159 | |||
BC172170.1 | - | 4 | 2141 |