Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172580.1 | NP_766168.1 | 3 | 1516 | ||
GenBank mRNA | AK014558.1 | - | 1390 | |||
AK039518.1 | - | 1014 | ||||
AY999300.1 | - | 3 | 1516 | |||
BC139306.1 | - | 3 | 1422 | 8862328 | ||
BC139328.1 | - | 3 | 1422 | 8862350 |