Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rd3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127685.1 | NP_001121157.1 | 3 | 615 | ||
GenBank mRNA | AK080710.1 | - | 2 | 1262 | ||
BC048943.1 | - | 4 | 871 | |||
BC147034.1 | - | 3 | 615 | 9056031 | ||
BC147035.1 | - | 3 | 615 | 9056043 | ||
BC147747.1 | - | 3 | 615 | 9056696 | ||
BC147749.1 | - | 3 | 615 | 9056698 |