Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam120a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033268.2 | NP_001028440.2 | 13 | 2398 | ||
GenBank mRNA | AK035074.1 | - | 3 | 313 | ||
AK172909.1 | - | 13 | 2587 | |||
BC010304.1 | - | 9 | 1466 | |||
BC033412.1 | - | 5 | 726 | |||
BC042582.1 | - | 12 | 1914 | |||
BC158134.1 | - | 13 | 2390 |