Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033272.1 | NP_001028444.1 | 13 | 4760 | ||
GenBank mRNA | AK151394.1 | - | 10 | 2642 | ||
AK158193.1 | - | 5 | 2294 | |||
AK220313.1 | - | 10 | 2482 | |||
BC035942.1 | - | 11 | 2778 | |||
BC040408.1 | - | 3 | 1344 | |||
BC040810.1 | - | 3 | 1344 | |||
BC138455.1 | - | 13 | 4717 | 8861475 | ||
BC145285.1 | - | 13 | 4717 | 9053839 |