Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104569.1 | NP_001098039.1 | 3 | 1268 | ||
GenBank mRNA | AF011416.1 | - | 1 | 673 |
Gene Symbol: Vmn2r104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF011417.1 | - | 673 |
Set Membership: |
G-protein coupled receptor |