Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080769.1 | NP_001074238.1 | 21 | 7471 | ||
GenBank mRNA | AK165438.1 | - | 3 | 3209 | ||
AK220293.1 | - | 21 | 7470 |