Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080769.1 | NP_001074238.1 | 7 | 932 | ||
GenBank mRNA | AK089790.1 | - | 7 | 950 | ||
AK220293.1 | - | 7 | 932 |