Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290345.1 | NP_001277274.1 | 34 | 3261 | ||
GenBank mRNA | AK079249.1 | - | 2 | 589 | ||
BC005781.1 | - | 15 | 1721 | |||
BC006037.1 | - | 19 | 2886 | |||
BC048923.1 | - | 2 | 1481 |