Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153774.1 | NP_722469.1 | 22 | 2970 | ||
GenBank mRNA | AF273672.1 | - | 22 | 2918 | ||
AF273673.1 | - | 22 | 2918 | |||
AJ309238.1 | - | 22 | 2912 | |||
AK036687.1 | - | 22 | 2920 | |||
BC056469.1 | - | 23 | 3118 | |||
BC079546.1 | - | 24 | 3085 | |||
BC098508.1 | - | 22 | 2920 | 30015513 |