Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178038.1 | NP_001171509.1 | 5 | 805 | ||
GenBank mRNA | BC051402.1 | - | 6 | 892 |