Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033285.1 | NP_001028457.1 | 19, 20 | 2760 | ||
GenBank mRNA | AF492452.1 | - | 5, 6 | 534 | ||
AK044466.1 | - | 1, 2 | 166 | |||
AK047837.1 | - | 3, 4 | 2522 | |||
BC158017.1 | - | 19, 20 | 2760 | 9007323 | ||
BC158095.1 | - | 19, 20 | 2760 | 9007382 |