Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033285.1 | NP_001028457.1 | 13 | 1862 | ||
GenBank mRNA | BC158017.1 | - | 13 | 1862 | 9007323 | |
BC158095.1 | - | 13 | 1862 | 9007382 |