Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081436.2 | NP_001074905.1 | 12 | 10871 | ||
NM_001114609.1 | NP_001108081.1 | 12 | 10962 | |||
GenBank mRNA | AK078621.1 | - | 1 | 2094 |