Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033291.2 | NP_001028463.1 | 2 | 196 | ||
NM_001198573.1 | NP_001185502.1 | 2 | 196 | |||
GenBank mRNA | AK030732.1 | - | 1 | 869 | ||
AK045613.1 | - | 2 | 196 | |||
AK050239.1 | - | 2 | 193 | |||
AK132295.1 | - | 2 | 196 | |||
AK166303.1 | - | 2 | 173 | |||
BC030843.1 | - | 2 | 145 | |||
BC117957.1 | - | 2 | 179 | 40098174 | ||
BC117958.1 | - | 2 | 179 | 40098175 |