Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100455.2 | NP_001093925.1 | 2, 3 | 258 | ||
GenBank mRNA | AY301267.1 | - | 2, 3 | 258 | ||
BE864901.1 | - | 1, 2 | 76 |