Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113184.1 | NP_001106655.1 | 5 | 981 | ||
NM_001113185.1 | NP_001106656.1 | 5 | 981 | |||
NM_017263.2 | NP_058959.2 | 5 | 981 | |||
GenBank mRNA | BC093608.1 | - | 5 | 953 | ||
M36421.1 | - | 4 | 531 | |||
M38063.1 | - | 4 | 531 | |||
M85037.1 | - | 5 | 692 |