Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081092.1 | NP_001074561.1 | 3 | 1414 | ||
GenBank mRNA | AY038601.1 | - | 3 | 270 |