Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ampd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033303.2 | NP_001028475.2 | 16 | 2259 | ||
GenBank mRNA | AK132524.1 | - | 16 | 2250 | ||
AK137549.1 | - | 14 | 2836 |