Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080818.2 | NP_001074287.1 | 16 | 4412 | ||
NM_001173553.1 | NP_001167024.1 | 14 | 4265 | |||
GenBank mRNA | AK142294.1 | - | 16 | 4415 | ||
BC072644.1 | - | 14 | 4220 | 30356962 |