Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080818.2 | NP_001074287.1 | 16 | 3287 | ||
NM_001173553.1 | NP_001167024.1 | 14 | 3140 | |||
GenBank mRNA | AK036556.1 | - | 13 | 2528 | ||
AK043956.1 | - | 16 | 3273 | |||
AK142294.1 | - | 16 | 3290 | |||
BC072644.1 | - | 14 | 3094 | 30356962 |