Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033306.1 | NP_001028478.1 | 6 | 2318 | ||
GenBank mRNA | BC028832.1 | - | 5 | 1180 | ||
BC048850.1 | - | 5 | 1185 | |||
BC058986.1 | - | 6 | 1820 |