Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183300.2 | NP_899123.2 | 3 | 323 | ||
GenBank mRNA | DQ303413.1 | - | 3 | 323 |