Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017276.2 | NP_058972.2 | 3 | 313 | ||
GenBank mRNA | BC061767.1 | - | 3 | 313 | 5621162 | |
X74401.1 | - | 3 | 243 |