Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AADACL3 SIRNA Assays ›
Gene Symbol:
Aadacl3
Gene Name:
arylacetamide deacetylase-like 3
Gene Aliases:
Gm435
Chromosome Location:
Chr.4: 144453771 - 144463756 on Build GRCm38
UniGene ID:
Mm.457844
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
502263
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Aadacl3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001085503.2
NP_001078972.1
4
2622
Back To Top
More Information
Panther Classification:
Molecular Function -
deacetylase
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
metabolic process
molecular_function
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
carboxylic ester hydrolase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline