Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam193a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243123.1 | NP_001230052.1 | 18 | 3568 | ||
GenBank mRNA | AK132046.1 | - | 2 | 207 | ||
BC037112.1 | - | 15 | 2705 | |||
BC099925.1 | - | 15 | 2605 | 30653351 |