Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A830010M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007574.2 | NP_001007575.2 | 6 | 3062 | ||
NM_001168557.1 | NP_001162029.1 | 6 | 1093 | |||
GenBank mRNA | AK048093.1 | - | 1 | 220 | ||
AK122444.1 | - | 6 | 3011 | |||
AK169666.1 | - | 6 | 1094 |
Gene Symbol: 1700028K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037115.1 | - | 1253 |