Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033877.4 | NP_001029049.2 | 4 | 860 | ||
GenBank mRNA | AK045148.1 | - | 5 | 981 | ||
AK053131.1 | - | 4 | 861 | |||
BC141170.1 | - | 4 | 812 |