Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dchs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162943.1 | NP_001156415.1 | 21 | 8204 | ||
GenBank mRNA | BC060096.1 | - | 2 | 704 |