Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033040.3 | NP_001028212.1 | 1 | 147 | ||
GenBank mRNA | AK140121.1 | - | 1 | 148 |