Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129787.1 | NP_001123259.1 | 18 | 2765 | ||
GenBank mRNA | AK220364.1 | - | 6 | 814 |