Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161362.3 | NP_001154834.1 | 2 | 1044 | ||
GenBank mRNA | AK042943.1 | - | 2 | 1008 | ||
AK076163.1 | - | 1 | 91 | |||
BC042842.1 | - | 1 | 479 | |||
BC151170.1 | - | 1 | 871 | 9088059 | ||
BC151171.1 | - | 1 | 866 | 9088060 |