Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161362.3 | NP_001154834.1 | 3 | 2827 | ||
NM_172144.3 | NP_742156.2 | 2 | 667 | |||
GenBank mRNA | AK035077.1 | - | 2 | 447 | ||
AK076163.1 | - | 2 | 1879 | |||
AK132175.1 | - | 2 | 668 | |||
AK169510.1 | - | 2 | 600 | |||
BC151170.1 | - | 2 | 2654 | 9088059 | ||
BC151171.1 | - | 2 | 2649 | 9088060 |