Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081127.1 | NP_001074596.1 | 2 | 308 | ||
GenBank mRNA | AK145103.1 | - | 2 | 310 | ||
BC157953.1 | - | 2 | 143 | 9007283 | ||
BC158014.1 | - | 2 | 141 | 9088151 | ||
BC172106.1 | - | 2 | 143 |