Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033335.3 | NP_001028507.2 | 6 | 1673 | ||
NM_001168294.1 | NP_001161766.1 | 6 | 1714 | |||
NM_001168295.1 | NP_001161767.1 | 5 | 1502 | |||
GenBank mRNA | AK138954.1 | - | 6 | 1674 | ||
BC049975.1 | - | 5 | 1492 | |||
BC139132.1 | - | 5 | 1562 | 8862154 | ||
BC139133.1 | - | 5 | 1562 | 8862155 |