Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044308.2 | NP_001037773.2 | 9 | 1736 | ||
GenBank mRNA | AK147585.1 | - | 9 | 1736 |