Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183046.1 | NP_898867.1 | 30 | 5169 | ||
GenBank mRNA | AK145779.1 | - | 16 | 3154 | ||
AK162252.1 | - | 11 | 1352 | |||
BC036147.1 | - | 12 | 2591 | |||
BC048954.1 | - | 11 | 1402 | |||
BC151061.1 | - | 30 | 5015 | 9087974 | ||
BC172148.1 | - | 30 | 5015 | 9054818 |