Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033344.3 | NP_001028516.2 | 6 | 3851 | ||
NM_001160049.1 | NP_001153521.1 | 6 | 3847 | |||
GenBank mRNA | AK142263.1 | - | 6 | 3852 | ||
BC059034.1 | - | 2 | 3200 |