Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081033.1 | NP_001074502.1 | 16, 17 | 2470 | ||
GenBank mRNA | AK042383.1 | - | 13, 14 | 1248 | ||
BC158125.1 | - | 16, 17 | 2610 | 9088197 |