Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033348.3 | NP_001028520.2 | 41 | 6320 | ||
GenBank mRNA | AK173140.1 | - | 9 | 1887 | ||
BC096528.1 | - | 13 | 2518 |