Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033348.3 | NP_001028520.2 | 15 | 2011 | ||
GenBank mRNA | AK038838.1 | - | 15 | 1919 | ||
CK638421.1 | - | 2 | 173 |