Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017240.2 | NP_058936.1 | 24 | 3265 | ||
XM_006251951.2 | XP_006252013.1 | 26 | 3415 | |||
GenBank mRNA | AB033760.1 | - | 3 | 185 | ||
X15939.1 | - | 24 | 3267 |