Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287224.1 | NP_001274153.1 | 2 | 546 | ||
GenBank mRNA | BB628321.1 | - | 1 | 141 | ||
KF954709.1 | - | 2 | 546 |