Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081358.2 | NP_001074827.1 | 11 | 2034 | ||
NM_001291452.1 | NP_001278381.1 | 11 | 2034 | |||
NM_001291453.1 | NP_001278382.1 | 11 | 2034 | |||
GenBank mRNA | AK122498.1 | - | 11 | 1169 | ||
AK141618.1 | - | 11 | 2034 | |||
BC138450.1 | - | 11 | 1941 | 8861470 |