Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085508.2 | NP_001078977.1 | 13 | 2230 | ||
GenBank mRNA | AK029713.1 | - | 13 | 2231 | ||
AK148848.1 | - | 2 | 159 | |||
BC147569.1 | - | 13 | 2101 | 9056559 | ||
BC147574.1 | - | 13 | 2101 | 9056564 | ||
BC151067.1 | - | 12 | 1832 | 9087980 | ||
BC151070.1 | - | 13 | 2101 | 9087983 |