Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gareml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167879.1 | NP_001161351.1 | 6 | 2874 | ||
GenBank mRNA | BC060268.1 | - | 5 | 2658 | ||
BC113782.1 | - | 4 | 2712 | |||
BC113783.1 | - | 4 | 2713 |