Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326475.1 | NP_001313404.1 | 2 | 484 | ||
NM_001326477.1 | NP_001313406.1 | 2 | 544 | |||
NM_004054.3 | NP_004045.1 | 2 | 487 | |||
GenBank mRNA | AK223388.1 | - | 2 | 397 | ||
AK290242.1 | - | 2 | 375 | |||
AK312458.1 | - | 2 | 375 | |||
BC020742.1 | - | 2 | 375 | 4690283 | ||
DA351375.1 | - | 2 | 488 | |||
DB013722.1 | - | 2 | 410 | |||
DB274838.1 | - | 2 | 372 | |||
U28488.1 | - | 2 | 364 | |||
U62027.1 | - | 2 | 435 | |||
Z73157.1 | - | 1 | 283 |
Set Membership: |
G-protein coupled receptor |